3,5-Bis(trifluoromethyl)benzyl chloride 98%

Supplier: Ambeed
Danger

A262318-5G A262318-25G
77113-710EA 9.27 USD
77113-710 77571-618
3,5-Bis(trifluoromethyl)benzyl chloride 98%
3,5-Bis(trifluoromethyl)benzyl chloride

Precaución: for Research Use Only

Formula: C₉H₅ClF₆
MDL Number: MFCD00009906
CAS Number: 75462-59-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR