Boc-3-chloro-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Phe(3-Cl)-OH

A262242-1G A262242-250MG A262242-5G
77113-704EA 10.31 USD
77113-704 77113-706 77685-746
Boc-3-chloro-D-phenylalanine 97%
Boc-3-chloro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₄
MW: 299.76 g/mol
MDL Number: MFCD00672516
CAS Number: 80102-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR