Guanidinium thiocyanate 99%

Supplier: Ambeed
Danger

A262103-100G A262103-25G
77113-700EA 18.53 USD
77113-700 77113-702
Guanidinium thiocyanate 99%
Guanidinium thiocyanate

Precaución: for Research Use Only

Formula: CH₅N₃·CHNS
MDL Number: MFCD00013027
CAS Number: 593-84-0
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR