α,α,α,3,4-Pentafluorotoluene 96%

Supplier: Ambeed
Danger

Synonyms: 3,4-Difluorobenzotrifluoride, 1,2-Difluoro-4-(trifluoromethyl)benzene

A261107-1G A261107-25G A261107-5G
77113-692EA 13.8 USD
77113-692 77113-694 77113-696
α,α,α,3,4-Pentafluorotoluene 96%
α,α,α,3,4-Pentafluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃F₅
MW: 182.09 g/mol
MDL Number: MFCD00077510
CAS Number: 32137-19-2
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR