NH125 99+%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-1-hexadecyl-2-methyl-1H-imidazol-3-ium iodide

A261079-10MG A261079-1MG A261079-50MG
77113-686EA 30.9 USD
77113-686 77113-688 77113-690
NH125 99+%
NH125

Precaución: for Research Use Only

Formula: C₂₇H₄₅IN₂
MW: 524.56 g/mol
MDL Number: MFCD07370143
CAS Number: 278603-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR