6-Methoxy-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxyindazole

A260817-1G A260817-250MG A260817-25G A260817-5G
77113-672EA 68.96 USD
77113-672 77113-674 77113-676 77113-678
6-Methoxy-1H-indazole 97%
6-Methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
MDL Number: MFCD09261132
CAS Number: 3522-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR