2-(Benzyloxy)-1,3,5-tribromobenzene 95%

Supplier: Ambeed
Warning

A258917-100MG A258917-250MG
77113-648EA 7.43 USD
77113-648 77113-650
2-(Benzyloxy)-1,3,5-tribromobenzene 95%
2-(Benzyloxy)-1,3,5-tribromobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉Br₃O
MW: 420.92 g/mol
MDL Number: MFCD00093101
CAS Number: 88486-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR