1-Bromo-3-methoxynaphthalene 95%

Supplier: Ambeed
Warning

A258746-100MG A258746-1G A258746-250MG A258746-5G
77113-640EA 42.44 USD
77113-640 77113-642 77113-644 77113-646
1-Bromo-3-methoxynaphthalene 95%
1-Bromo-3-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.09 g/mol
MDL Number: MFCD18410560
CAS Number: 5111-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR