2-Hydroxy-6-methylnicotinonitrile 97%

Supplier: Ambeed
Warning

A258686-100G A258686-10G A258686-1G A258686-250MG A258686-25G A258686-5G
77113-628EA 282.2 USD
77113-628 77113-630 77113-632 77113-634 77113-636 77113-638
2-Hydroxy-6-methylnicotinonitrile 97%
2-Hydroxy-6-methylnicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD00006013
CAS Number: 4241-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR