Domiphen bromide 98%

Supplier: Ambeed
Warning

Synonyms: (Dodecyldimethyl-2-phenoxyethyl)ammonium bromide, N,N-Dimethyl-N-(2-phenoxyethyl)dodecan-1-aminium bromide

A258344-1000G A258344-100G A258344-25G A258344-500G
77113-610EA 183.35 USD
77113-610 77113-612 77113-614 77113-616
Domiphen bromide 98%
Domiphen bromide

Precaución: for Research Use Only

Formula: C₂₂H₄₀BrNO
MW: 414.48 g/mol
MDL Number: MFCD00011769
CAS Number: 538-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR