tert-Butyl-4-(2-aminoethyl)-4-hydroxypiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(2-aminoethyl)-4-hydroxypiperidine-1-carboxylate

A258013-1G A258013-250MG
77113-602EA 239.77 USD
77113-602 77113-604
tert-Butyl-4-(2-aminoethyl)-4-hydroxypiperidine-1-carboxylate 95%
tert-Butyl-4-(2-aminoethyl)-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₃
MW: 244.3306 g/mol
MDL Number: MFCD11111570
CAS Number: 1179338-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR