2-Boc-aminothiazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Boc-Aminothiazole-5-carboxylic acid, 2-(tert-Butoxycarbonylamino)thiazole-5-carboxylic acid, 2-(Boc-amino)thiazole-5-carboxylic acid

A257668-100G A257668-10G A257668-1G A257668-250MG A257668-25G A257668-5G
77113-580EA 649.28 USD
77113-580 77113-582 77113-584 77113-586 77113-588 77113-590
2-Boc-aminothiazole-5-carboxylic acid 97%
2-Boc-aminothiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄S
MW: 244.2676 g/mol
MDL Number: MFCD06796614
CAS Number: 302964-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR