Boc-(R)-3-amino-5-phenylpentanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-5-phenylpentanoic acid

A256841-100MG A256841-1G A256841-250MG
77113-572EA 25.47 USD
77113-572 77113-574 77113-576
Boc-(R)-3-amino-5-phenylpentanoic acid 98%
Boc-(R)-3-amino-5-phenylpentanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₄
MW: 293.37 g/mol
MDL Number: MFCD01076262
CAS Number: 218608-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR