2-Amino-5-bromo-4-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-4-methylpyridin-2-amine, 5-Bromo-4-methyl-2-pyridylamine, 2-Amino-5-bromo-4-picoline
A256139-100G
A256139-10G
A256139-1G
A256139-25G
A256139-500G
A256139-5G
77113-552EA
51.51
USD
77113-552
77113-554
77113-556
77113-558
77113-560
77113-562
2-Amino-5-bromo-4-methylpyridine 98%
2-Amino-5-bromo-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₆H₇BrN₂ MW: 187.04 g/mol |
MDL Number:
MFCD03427660 CAS Number: 98198-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...