2-Amino-5-chloro-4-methylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-chloro-4-picoline

A255675-10G A255675-1G A255675-5G
77113-546EA 15.45 USD
77113-546 77113-548 77113-550
2-Amino-5-chloro-4-methylpyridine 95%
2-Amino-5-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.5900 g/mol
MDL Number: MFCD06410759
CAS Number: 36936-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR