1,1'-Bis(4-boronobenzyl)-[4,4'-bipyridine]-1,1'-diium bromide 95%

Supplier: Ambeed
Warning

A255614-100MG A255614-1G A255614-250MG
77113-540EA 89.61 USD
77113-540 77113-542 77113-544
1,1'-Bis(4-boronobenzyl)-[4,4'-bipyridine]-1,1'-diium bromide 95%
1,1'-Bis(4-boronobenzyl)-[4,4'-bipyridine]-1,1'-diium bromide

Precaución: for Research Use Only

Formula: C₂₄H₂₄B₂N₂O₄·2Br
MDL Number: MFCD24849237
CAS Number: 436853-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR