N-Acetyl-DL-tryptophan 98%

Supplier: Ambeed
Warning

A254375-100G A254375-25G
77113-520EA 72.1 USD
77113-520 77113-522 77113-524 77113-526
N-Acetyl-DL-tryptophan 98%
N-Acetyl-DL-tryptophan

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₃
MDL Number: MFCD00005644
CAS Number: 87-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR