6-Bromoindoline hydrochloride 95%

Supplier: Ambeed
Warning

A254007-1G A254007-250MG
77113-516EA 85.94 USD
77113-516 77113-518
6-Bromoindoline hydrochloride 95%
6-Bromoindoline hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉BrClN
MW: 234.52 g/mol
MDL Number: MFCD09026836
CAS Number: 1187933-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR