Tie2 Kinase Inhibitor 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(6-Methoxynaphthalen-2-yl)-2-(4-(methylsulphinyl)phenyl)-1H-imidazol-5-yl)pyridine, 4-[4-(6-methoxy-2-naphthalenyl)-2-[4-(methylsulphinyl)phenyl]-1H-imidazol-5-yl]-pyridine, Tie2 kinase inhibitor 1, 4-(6-Methoxy-2-naphthyl)-2-(4-methylsulphinylphenyl)-5-(4-pyridyl)-1H-imidazole

A253059-100MG A253059-10MG A253059-1MG A253059-50MG
77113-504EA 306.61 USD
77113-504 77113-506 77113-508 77113-510
Tie2 Kinase Inhibitor 98%
Tie2 Kinase Inhibitor

Precaución: for Research Use Only

Formula: C₂₆H₂₁N₃O₂S
MW: 439.5 g/mol
MDL Number: MFCD18384977
CAS Number: 948557-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR