N6-(2-((5-(Diethylamino)pentan-2-yl)amino)-6-methylpyrimidin-4-yl)-2-methylquinoline-4,6-diamine trihydrochloride 98%
Supplier: Ambeed
|
Warning
|
A252594-10MG
A252594-1MG
A252594-50MG
A252594-5MG
77113-496EA
100.79
USD
77113-496
77113-498
77113-500
77113-502
N6-(2-((5-(Diethylamino)pentan-2-yl)amino)-6-methylpyrimidin-4-yl)-2-methylquinoline-4,6-diamine trihydrochloride 98%
N6-(2-((5-(Diethylamino)pentan-2-yl)amino)-6-methylpyrimidin-4-yl)-2-methylquinoline-4,6-diamine trihydrochloride
Precaución: for Research Use Only
Formula:
C₂₄H₃₈Cl₃N₇ MW: 530.96 g/mol |
MDL Number:
MFCD08703096 CAS Number: 1177865-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...