m-Tolylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Methylphenyl)hydrazine hydrochloride, 3-Tolylhydrazine hydrochloride, m-Tolylhydrazine hydrochloride, m-Tolylhydraziniumchloride, 3-Methylphenylhydrazine hydrochloride, 1-(3-Tolyl)hydrazine hydrochloride, 3-Methylphenylhydrazinium chloride

A252549-10G A252549-1G A252549-25G A252549-5G
77113-488EA 11.33 USD
77113-488 77113-490 77113-492 77113-494
m-Tolylhydrazine hydrochloride 97%
m-Tolylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀N₂·HCl
MW: 158.63 g/mol
MDL Number: MFCD00012932
CAS Number: 637-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR