5-Bromo-2-isopropoxypyridine 97%

Supplier: Ambeed
Warning

A252192-1G A252192-250MG A252192-5G
77113-480EA 27.59 USD
77113-480 77113-482 77685-716
5-Bromo-2-isopropoxypyridine 97%
5-Bromo-2-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD07368869
CAS Number: 870521-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR