Glycyl-D-leucine 99%

Supplier: Ambeed
Warning

Synonyms: H-Gly-D-Leu-OH, (R)-2-(2-Aminoacetamido)-4-methylpentanoic acid

A251964-100G A251964-1G A251964-25G A251964-5G
77113-472EA 1070.45 USD
77113-472 77113-474 77113-476 77113-478
Glycyl-D-leucine 99%
Glycyl-D-leucine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₃
MW: 188.23 g/mol
MDL Number: MFCD00066050
CAS Number: 688-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR