4,6-Dichlorosalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-6-hydroxybenzaldehyde

A250664-1G A250664-5G
77113-448EA 58.35 USD
77113-448 77113-450
4,6-Dichlorosalicylaldehyde 98%
4,6-Dichlorosalicylaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
MDL Number: MFCD00016590
CAS Number: 78443-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR