(1R)-1-(3-Fluorophenyl)ethylamine 95%

Supplier: Ambeed
Danger

A250134-1G A250134-250MG A250134-5G
77113-430EA 30.9 USD
77113-430 77113-432 77113-434
(1R)-1-(3-Fluorophenyl)ethylamine 95%
(1R)-1-(3-Fluorophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MDL Number: MFCD06761859
CAS Number: 761390-58-3
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR