5-Fluoro-4-iodo-2-methoxypyridine 98%

Supplier: Ambeed
Warning

A249748-100MG A249748-1G A249748-250MG A249748-5G A249748-25G
77113-422EA 28.65 USD
77113-422 77113-424 77113-426 77113-428 77568-428
5-Fluoro-4-iodo-2-methoxypyridine 98%
5-Fluoro-4-iodo-2-methoxypyridine

Precaución: For research use only.

Formula: C₆H₅FINO
MW: 253.013 g/mol
MDL Number: MFCD06659503
CAS Number: 884495-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR