3-Methylpyrrole 97%

Supplier: Ambeed
Danger

Synonyms: 3-Methyl-1H-pyrrole

A249384-5G A249384-1G
77113-420EA 360.71 USD
77113-420 77568-418
3-Methylpyrrole 97%
3-Methylpyrrole

Precaución: For research use only.

Formula: C₅H₇N
MW: 81.12 g/mol
MDL Number: MFCD00083419
CAS Number: 616-43-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR