4-Bromo-2-chlorobenzylamine 95%

Supplier: Ambeed
Danger

A248867-1G A248867-250MG
77113-410EA 307.97 USD
77113-410 77113-412
4-Bromo-2-chlorobenzylamine 95%
4-Bromo-2-chlorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇BrClN
MW: 220.49 g/mol
MDL Number: MFCD06213844
CAS Number: 771574-32-4
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR