Boc-S-4-methylbenzyl-D-cysteine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Cys(Mbzl)-OH, Boc-D-Cys(pMeBzl)-OH

A248834-1G A248834-5G
77113-406EA 46.68 USD
77113-406 77113-408
Boc-S-4-methylbenzyl-D-cysteine 97%
Boc-S-4-methylbenzyl-D-cysteine

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₄S
MW: 325.423 g/mol
MDL Number: MFCD00076924
CAS Number: 61925-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR