3-Fluorophthalonitrile 95%

Supplier: Ambeed
Warning

A248671-1G A248671-250MG A248671-5G
77113-394EA 28.85 USD
77113-394 77113-396 77113-398
3-Fluorophthalonitrile 95%
3-Fluorophthalonitrile

Precaución: for Research Use Only

Formula: C8H3FN2
MDL Number: MFCD00052458
CAS Number: 65610-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR