7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine 95%

Supplier: Ambeed
Warning

A248091-100MG A248091-10MG A248091-1MG A248091-50MG A248091-5MG
77113-380EA 200.52 USD
77113-380 77113-382 77113-384 77113-386 77113-388
7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine 95%
7-Methoxy-4-methyl-2,3,4,5-tetrahydrobenzo[f][1,4]thiazepine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NOS
MW: 209.30 g/mol
MDL Number: MFCD12546447
CAS Number: 927871-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR