2,6-Dichloro-4-nitrotoluene 97%
Supplier: Ambeed
|
Warning
|
A247288-1G
A247288-250MG
A247288-5G
77113-368EA
13.39
USD
77113-368
77113-370
77685-697
2,6-Dichloro-4-nitrotoluene 97%
2,6-Dichloro-4-nitrotoluene
Precaución: for Research Use Only
Formula:
C₇H₅Cl₂NO₂ |
MDL Number:
MFCD00174267 CAS Number: 7149-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...