(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 98%

Supplier: Ambeed
Danger

Synonyms: (S)-(1-Ethylpyrrolidin-2-yl)methanamine, (S)-2-Aminomethyl-1-ethylpyrrolidine, (S)-1-ethyl-2-aminomethylpyrrolidine

A246855-1G A246855-25G A246855-5G
77113-356EA 6.37 USD
77113-356 77113-358 77113-360
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 98%
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD00191371
CAS Number: 22795-99-9
UN: 2733
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR