(2-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A245633-100MG A245633-1G A245633-250MG
77113-344EA 36.08 USD
77113-344 77113-346 77113-348
(2-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid 98%
(2-((4-(Trifluoromethoxy)phenoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂BF₃O₄
MW: 312.05 g/mol
MDL Number: MFCD07369751
CAS Number: 849062-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR