2,4-Dihydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A245052-100G A245052-10G A245052-25G A245052-500G A245052-5G A245052-1000G
77113-322EA 22.28 USD
77113-322 77113-324 77113-326 77113-328 77113-330 77685-687
2,4-Dihydroxybenzaldehyde 98%
2,4-Dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
MDL Number: MFCD00011686
CAS Number: 95-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR