5-Iodo-2-methoxypyridine-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Iodo-2-methoxynicotinic acid
A245037-100MG
A245037-250MG
A245037-5G
77113-316EA
13.39
USD
77113-316
77113-318
77113-320
5-Iodo-2-methoxypyridine-3-carboxylic acid 95%
5-Iodo-2-methoxypyridine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₆INO₃ |
MDL Number:
MFCD22573540 CAS Number: 1407521-93-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...