3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid 96%

Supplier: Ambeed
Warning

A244354-1G A244354-250MG A244354-25G A244354-5G
77113-300EA 13.8 USD
77113-300 77113-302 77113-304 77113-306
3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid 96%
3-Amino-5-(4-chlorophenyl)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C11H8ClNO2S
MW: 253.71 g/mol
MDL Number: MFCD09909681
CAS Number: 187949-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR