N,N-Dimethylpropionamide 97%

Supplier: Ambeed
Warning

A241987-100G A241987-10G A241987-5G A241987-25G A241987-500G A241987-1000G
77113-260EA 20.16 USD
77113-260 77113-262 77113-264 77304-240 77568-352 77685-677
N,N-Dimethylpropionamide 97%
N,N-Dimethylpropionamide

Precaución: For research use only.

Formula: C₅H₁₁NO
MW: 101.15 g/mol
MDL Number: MFCD00009301
CAS Number: 758-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR