4-Chloro-N-(cyclopropylmethyl)aniline hydrochloride 97%

Supplier: Ambeed
Warning

A240657-1G A240657-250MG
77113-250EA 44.56 USD
77113-250 77113-252
4-Chloro-N-(cyclopropylmethyl)aniline hydrochloride 97%
4-Chloro-N-(cyclopropylmethyl)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃Cl₂N
MW: 218.12 g/mol
MDL Number: MFCD09026755
CAS Number: 1785761-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR