1-Methyl-1H-pyrazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A239339-100G A239339-10G A239339-1G A239339-250MG A239339-25G A239339-5G
77113-214EA 114.34 USD
77113-214 77113-216 77113-218 77113-220 77113-222 77113-224
1-Methyl-1H-pyrazole-4-carboxylic acid 95%
1-Methyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂
MDL Number: MFCD00159641
CAS Number: 5952-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR