(R)-(+)-3-Aminoquinuclidine dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Aminoquinuclidine dihydrochloride

A239225-1G A239225-250MG A239225-25G
77113-208EA 15.45 USD
77113-208 77113-210 77113-212
(R)-(+)-3-Aminoquinuclidine dihydrochloride 95%
(R)-(+)-3-Aminoquinuclidine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00191752
CAS Number: 123536-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR