D-Leucine methyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Leu-OMe.HCl

A238710-100G A238710-10G A238710-1G A238710-25G A238710-5G
77113-186EA 101.97 USD
77113-186 77113-188 77113-190 77113-192 77113-194
D-Leucine methyl ester hydrochloride 98%
D-Leucine methyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 181.66 g/mol
MDL Number: MFCD00066115
CAS Number: 5845-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR