4-Chloro-1,2-difluorobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 3,4-Difluorochlorobenzene

A238663-10G A238663-1G A238663-25G A238663-5G
77113-178EA 11.67 USD
77113-178 77113-180 77113-182 77113-184
4-Chloro-1,2-difluorobenzene 98%
4-Chloro-1,2-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃ClF₂
MW: 148.54 g/mol
MDL Number: MFCD00042572
CAS Number: 696-02-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR