Boc-D-THZ-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Thz-OH, Boc-D-thiazolidine-4-carboxylic acid

A237439-100G A237439-10G A237439-1G A237439-250MG A237439-25G A237439-5G
77113-152EA 419.21 USD
77113-152 77113-154 77113-156 77113-158 77113-160 77113-162
Boc-D-THZ-OH 95%
Boc-D-THZ-OH

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄S
MDL Number: MFCD00077001
CAS Number: 63091-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR