6-Chloro-3-fluoropicolinic acid 98%

Supplier: Ambeed
Warning

A236550-100G A236550-100MG A236550-10G A236550-1G A236550-250MG A236550-25G A236550-5G
77113-138EA 1071.2 USD
77113-138 77113-140 77113-142 77113-144 77113-146 77113-148 77113-150
6-Chloro-3-fluoropicolinic acid 98%
6-Chloro-3-fluoropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MW: 175.545 g/mol
MDL Number: MFCD04972387
CAS Number: 884494-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR