(3-Methoxyphenyl)hydrazine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Methoxyphenylhydrazine hydrochloride

A236316-100G A236316-10G A236316-25G A236316-500G A236316-5G
77113-128EA 167.63 USD
77113-128 77113-130 77113-132 77113-134 77113-136
(3-Methoxyphenyl)hydrazine hydrochloride 98%
(3-Methoxyphenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MW: 174.63 g/mol
MDL Number: MFCD00044416
CAS Number: 39232-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR