L(+)-Arginine 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH
A234349-100G
A234349-25G
A234349-500G
A234349-1000G
77113-100EA
6.18
USD
77113-100
77113-102
77113-104
77685-645
L(+)-Arginine 98+%
L(+)-Arginine
Precaución: for Research Use Only
Formula:
C₆H₁₄N₄O₂ MW: 174.2 g/mol Boiling Pt: 368 Melting Pt: 238 |
MDL Number:
MFCD00002635 CAS Number: 74-79-3 Merck Index: 13,00785 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...