4-Iodoaniline 98%

Supplier: Ambeed
Warning

A234175-1000G A234175-100G A234175-10G A234175-500G A234175-25G
77113-092EA 356.47 USD
77113-092 77113-094 77113-096 77113-098 77303-972
4-Iodoaniline 98%
4-Iodoaniline

Precaución: for Research Use Only

Formula: C₆H₆IN
MW: 219.02 g/mol
MDL Number: MFCD00007848
CAS Number: 540-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR