2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-phenyl-5-(trifluoromethyl)pyrazole

A233174-1G A233174-5G
77113-066EA 12.74 USD
77113-066 77113-068
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol 95%
2-Phenyl-5-trifluoromethyl-2H-pyrazol-3-ol

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂O
MW: 228.1700 g/mol
MDL Number: MFCD03714773
CAS Number: 96145-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR