2-Ethoxy-5-pyridineboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid

A232887-10G A232887-1G A232887-250MG A232887-100G A232887-5G
77113-056EA 33.95 USD
77113-056 77113-058 77113-060 77612-276 77685-644
2-Ethoxy-5-pyridineboronic acid 97%
2-Ethoxy-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
MDL Number: MFCD06201032
CAS Number: 612845-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR